Assay Details
| Gene Symbol: LOC4325486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757621.2 | XP_015613107.1 | 12 - 13 | 1432 | 62 | |
| XM_015757626.2 | XP_015613112.1 | 11 - 12 | 1925 | 62 | ||
| XM_015757635.2 | XP_015613121.1 | 12 - 13 | 1434 | 62 | ||
| GenBank mRNA | AK062266.1 | - | 5 - 6 | 645 | 62 | |
| AK065031.1 | - | 12 - 13 | 1429 | 62 | ||
More Information
Set Membership: |
|

LOC4325486
Genomic Map