Assay Details
Gene Symbol: LOC4325354 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772328.2 | XP_015627814.1 | 8 - 9 | 917 | 93 | |
GenBank mRNA | AK240913.1 | - | 8 - 9 | 924 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
