Assay Details
Gene Symbol: LOC4327063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766457.2 | XP_015621943.1 | 2 - 3 | 741 | 86 | |
GenBank mRNA | AK068290.1 | - | 2 - 3 | 725 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
