Assay Details
Gene Symbol: LOC4324278 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757095.1 | XP_015612581.1 | 14 - 15 | 3878 | 132 | |
GenBank mRNA | AK101990.1 | - | 3 - 4 | 1275 | 132 | |
AK106519.1 | - | 3 - 4 | 1205 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
