Assay Details
Gene Symbol: LOC4325161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766590.2 | XP_015622076.1 | 10 - 11 | 1459 | 119 | |
GenBank mRNA | AK065588.1 | - | 10 - 11 | 1321 | 119 | |
AK068440.1 | - | 4 - 5 | 531 | 119 | ||
EU703792.1 | - | 10 - 11 | 1114 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
