Assay Details
Gene Symbol: LOC4325161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766590.2 | XP_015622076.1 | 12 - 13 | 1614 | 83 | |
GenBank mRNA | AK061640.1 | - | 2 - 3 | 98 | 83 | |
AK065588.1 | - | 12 - 13 | 1476 | 83 | ||
AK068440.1 | - | 6 - 7 | 686 | 83 | ||
EU703792.1 | - | 12 - 13 | 1269 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
