Assay Details
Gene Symbol: LOC4325161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766590.2 | XP_015622076.1 | 3 - 4 | 674 | 103 | |
GenBank mRNA | AK065588.1 | - | 3 - 4 | 536 | 103 | |
EU703792.1 | - | 3 - 4 | 329 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
