Assay Details
Gene Symbol: LOC4325161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766590.2 | XP_015622076.1 | 8 - 9 | 1240 | 86 | |
GenBank mRNA | AK065588.1 | - | 8 - 9 | 1102 | 86 | |
AK068440.1 | - | 2 - 3 | 312 | 86 | ||
EU703792.1 | - | 8 - 9 | 895 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
