Assay Details
Gene Symbol: LOC4325161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766590.2 | XP_015622076.1 | 9 - 10 | 1349 | 78 | |
GenBank mRNA | AK065588.1 | - | 9 - 10 | 1211 | 78 | |
AK068440.1 | - | 3 - 4 | 421 | 78 | ||
EU703792.1 | - | 9 - 10 | 1004 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
