Assay Details
| Gene Symbol: LOC4327728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792836.2 | XP_015648322.1 | 10 - 11 | 2125 | 106 | |
| GenBank mRNA | AK100135.1 | - | 9 - 10 | 1444 | 106 | |
More Information
Set Membership: |
|

LOC4327728
Genomic Map