Assay Details
| Gene Symbol: LOC4327728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792836.2 | XP_015648322.1 | 17 - 18 | 3649 | 89 | |
| GenBank mRNA | AK058981.1 | - | 3 - 4 | 414 | 89 | |
| AK100135.1 | - | 16 - 17 | 2968 | 89 | ||
More Information
Set Membership: |
|

LOC4327728
Genomic Map