Assay Details
| Gene Symbol: LOC4327728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792836.2 | XP_015648322.1 | 7 - 8 | 1232 | 106 | |
| GenBank mRNA | AK100135.1 | - | 6 - 7 | 551 | 106 | |
More Information
Set Membership: |
|

LOC4327728
Genomic Map