Assay Details
Gene Symbol: LOC4325312 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015767352.2 | XP_015622838.1 | 2 - 3 | 233 | 84 | |
GenBank mRNA | AB015288.1 | - | 2 - 3 | 144 | 84 | |
AB015971.1 | - | 2 - 3 | 148 | 84 | ||
AK072731.1 | - | 2 - 3 | 167 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
