Assay Details
| Gene Symbol: LOC4326608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776785.2 | XP_015632271.1 | 10 - 11 | 1719 | 120 | |
| GenBank mRNA | AK073890.1 | - | 10 - 11 | 1725 | 120 | |
| AY693783.1 | - | 5 - 6 | 631 | 120 | ||
More Information
Set Membership: |
|

LOC4326608
Genomic Map