Assay Details
| Gene Symbol: LOC4326608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776785.2 | XP_015632271.1 | 12 - 13 | 1906 | 156 | |
| XM_015776864.2 | XP_015632350.1 | 10 - 11 | 1816 | 156 | ||
| GenBank mRNA | AK073890.1 | - | 12 - 13 | 1912 | 156 | |
| AY693783.1 | - | 7 - 8 | 818 | 156 | ||
More Information
Set Membership: |
|

LOC4326608
Genomic Map