Assay Details
| Gene Symbol: LOC4326608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776785.2 | XP_015632271.1 | 17 - 18 | 2437 | 103 | |
| XM_015776864.2 | XP_015632350.1 | 15 - 16 | 2347 | 103 | ||
| GenBank mRNA | AK073890.1 | - | 17 - 18 | 2443 | 103 | |
| AY693783.1 | - | 12 - 13 | 1349 | 103 | ||
| AY693789.1 | - | 2 - 3 | 231 | 103 | ||
More Information
Set Membership: |
|

LOC4326608
Genomic Map