Assay Details
| Gene Symbol: LOC4326608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776785.2 | XP_015632271.1 | 4 - 5 | 571 | 72 | |
| XM_015776864.2 | XP_015632350.1 | 4 - 5 | 667 | 72 | ||
| GenBank mRNA | AK073890.1 | - | 4 - 5 | 578 | 72 | |
More Information
Set Membership: |
|

LOC4326608
Genomic Map