Assay Details
Gene Symbol: LOC4327850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766789.2 | XP_015622275.1 | 1 - 2 | 1045 | 68 | |
GenBank mRNA | AK101958.1 | - | 1 - 2 | 969 | 68 | |
AK105412.1 | - | 1 - 2 | 1024 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
