Assay Details
| Gene Symbol: LOC4327860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015766422.2 | XP_015621908.1 | 3 - 4 | 1172 | 97 | |
| GenBank mRNA | AK071667.1 | - | 3 - 4 | 985 | 97 | |
More Information
Set Membership: |
|

LOC4327860
Genomic Map