Assay Details
Gene Symbol: LOC4324389 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015791666.2 | XP_015647152.1 | 1 - 2 | 343 | 94 | |
XM_015791675.2 | XP_015647161.1 | 1 - 2 | 343 | 94 | ||
GenBank mRNA | AK060234.1 | - | 325 | 94 | ||
AK100939.1 | - | 1 - 2 | 329 | 94 | ||
AK106904.1 | - | 1 - 2 | 328 | 94 | ||
AK120014.1 | - | 1 - 2 | 334 | 94 | ||
AK121420.1 | - | 1 - 2 | 335 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
