Assay Details
| Gene Symbol: LOC4324716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762683.2 | XP_015618169.1 | 14 - 15 | 1632 | 90 | |
| XM_015762690.2 | XP_015618176.1 | 14 - 15 | 1618 | 90 | ||
| XM_015762698.2 | XP_015618184.1 | 14 - 15 | 1630 | 90 | ||
| GenBank mRNA | AK120199.1 | - | 11 - 12 | 2198 | 90 | |
More Information
Set Membership: |
|

LOC4324716
Genomic Map