Assay Details
| Gene Symbol: LOC4324716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762683.2 | XP_015618169.1 | 1 - 2 | 176 | 60 | |
| XM_015762690.2 | XP_015618176.1 | 1 - 2 | 177 | 60 | ||
| XM_015762698.2 | XP_015618184.1 | 1 - 2 | 174 | 60 | ||
| GenBank mRNA | AK069524.1 | - | 1 - 2 | 177 | 60 | |
| AK120199.1 | - | 1 - 2 | 177 | 60 | ||
More Information
Set Membership: |
|

LOC4324716
Genomic Map