Assay Details
| Gene Symbol: LOC4324716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762683.2 | XP_015618169.1 | 8 - 9 | 891 | 138 | |
| XM_015762690.2 | XP_015618176.1 | 8 - 9 | 892 | 138 | ||
| XM_015762698.2 | XP_015618184.1 | 8 - 9 | 889 | 138 | ||
| GenBank mRNA | AK069524.1 | - | 5 - 6 | 1469 | 138 | |
| AK120199.1 | - | 5 - 6 | 1452 | 138 | ||
More Information
Set Membership: |
|

LOC4324716
Genomic Map