Assay Details
Gene Symbol: LOC4328064 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770411.1 | XP_015625897.1 | 4 - 5 | 1632 | 83 | |
GenBank mRNA | AK102050.1 | - | 4 - 5 | 1633 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
