Assay Details
| Gene Symbol: LOC4328127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770824.2 | XP_015626310.1 | 2 - 3 | 231 | 73 | |
| XM_015770825.2 | XP_015626311.1 | 2 - 3 | 273 | 73 | ||
| XM_015770826.2 | XP_015626312.1 | 2 - 3 | 225 | 73 | ||
| GenBank mRNA | AK121653.1 | - | 1 - 2 | 216 | 73 | |
More Information
Set Membership: |
|

LOC4328127
Genomic Map