Assay Details
Gene Symbol: LOC4328149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770214.2 | XP_015625700.1 | 1 - 2 | 263 | 75 | |
GenBank mRNA | AK061564.1 | - | 1 - 2 | 140 | 75 | |
AK061953.1 | - | 1 - 2 | 251 | 75 | ||
AK101060.1 | - | 1 - 2 | 262 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
