Assay Details
Gene Symbol: LOC4328149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770214.2 | XP_015625700.1 | 4 - 5 | 694 | 61 | |
GenBank mRNA | AK061564.1 | - | 4 - 5 | 571 | 61 | |
AK061953.1 | - | 4 - 5 | 682 | 61 | ||
AK101060.1 | - | 4 - 5 | 693 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
