Assay Details
| Gene Symbol: LOC4328199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769928.2 | XP_015625414.1 | 5 - 6 | 1132 | 100 | |
| XM_015769930.2 | XP_015625416.1 | 5 - 6 | 1260 | 100 | ||
| GenBank mRNA | AK103571.1 | - | 5 - 6 | 1127 | 100 | |
More Information
Set Membership: |
|

LOC4328199
Genomic Map