Assay Details
| Gene Symbol: LOC4328208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771225.2 | XP_015626711.1 | 6 - 7 | 1163 | 89 | |
| XM_015771226.2 | XP_015626712.1 | 6 - 7 | 1132 | 89 | ||
More Information
Set Membership: |
|

LOC4328208
Genomic Map