Assay Details
| Gene Symbol: LOC4328208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771225.2 | XP_015626711.1 | 9 - 10 | 1420 | 74 | |
| XM_015771226.2 | XP_015626712.1 | 9 - 10 | 1389 | 74 | ||
| GenBank mRNA | AK060180.1 | - | 5 - 6 | 429 | 74 | |
More Information
Set Membership: |
|

LOC4328208
Genomic Map