Assay Details
| Gene Symbol: LOC4328208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771225.2 | XP_015626711.1 | 12 - 13 | 1756 | 73 | |
| XM_015771226.2 | XP_015626712.1 | 12 - 13 | 1725 | 73 | ||
| GenBank mRNA | AK060180.1 | - | 8 - 9 | 765 | 73 | |
More Information
Set Membership: |
|

LOC4328208
Genomic Map