Assay Details
| Gene Symbol: LOC4328330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769957.2 | XP_015625443.1 | 1 - 2 | 193 | 114 | |
| GenBank mRNA | AK073943.1 | - | 1 - 2 | 173 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015769957.2 transcripts.
Set Membership: |
|

LOC4328330
Genomic Map