Assay Details
| Gene Symbol: LOC4328330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769957.2 | XP_015625443.1 | 6 - 7 | 753 | 86 | |
| GenBank mRNA | AK073943.1 | - | 6 - 7 | 733 | 86 | |
More Information
Set Membership: |
|

LOC4328330
Genomic Map