Assay Details
| Gene Symbol: LOC4328715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770253.1 | XP_015625739.1 | 1 - 2 | 254 | 79 | |
| GenBank mRNA | AK107283.1 | - | 1 - 2 | 239 | 79 | |
| EF122479.1 | - | 1 - 2 | 222 | 79 | ||
More Information
Set Membership: |
|

LOC4328715
Genomic Map