Assay Details
Gene Symbol: LOC4328877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771644.2 | XP_015627130.1 | 1 - 2 | 349 | 76 | |
GenBank mRNA | AK243018.1 | - | 1 - 2 | 333 | 76 | |
AY429650.1 | - | 1 - 2 | 338 | 76 | ||
EF122464.1 | - | 1 - 2 | 322 | 76 | ||
JQ694716.1 | - | 1 - 2 | 271 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
