Assay Details
Gene Symbol: LOC4329094 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768439.2 | XP_015623925.1 | 3 - 4 | 2328 | 113 | |
GenBank mRNA | AK121646.1 | - | 3 - 4 | 2327 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
