Assay Details
| Gene Symbol: LOC4329234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771614.2 | XP_015627100.1 | 7 - 8 | 1438 | 83 | |
| GenBank mRNA | AK062153.1 | - | 6 - 7 | 785 | 83 | |
| AK067100.1 | - | 7 - 8 | 1429 | 83 | ||
More Information
Set Membership: |
|

LOC4329234
Genomic Map