Assay Details
| Gene Symbol: LOC4329407 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771372.2 | XP_015626858.1 | 10 - 11 | 2806 | 140 | |
| GenBank mRNA | AK071406.1 | - | 10 - 11 | 2762 | 140 | |
More Information
Set Membership: |
|

LOC4329407
Genomic Map