Assay Details
Gene Symbol: LOC4329631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771918.2 | XP_015627404.1 | 2 - 3 | 500 | 93 | |
GenBank mRNA | AJ428900.1 | - | 1 - 2 | 171 | 93 | |
AK068448.1 | - | 2 - 3 | 442 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
