Assay Details
Gene Symbol: LOC4329631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015771918.2 | XP_015627404.1 | 4 - 5 | 965 | 109 | |
GenBank mRNA | AJ428900.1 | - | 3 - 4 | 636 | 109 | |
AK068448.1 | - | 4 - 5 | 907 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
