Assay Details
| Gene Symbol: LOC4329652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770565.2 | XP_015626051.1 | 1 - 2 | 541 | 71 | |
| GenBank mRNA | AK072252.1 | - | 1 - 2 | 384 | 71 | |
| AK073161.1 | - | 1 - 2 | 237 | 71 | ||
More Information
Set Membership: |
|

LOC4329652
Genomic Map