Assay Details
| Gene Symbol: LOC4329652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770565.2 | XP_015626051.1 | 2 - 3 | 879 | 65 | |
| GenBank mRNA | AK072252.1 | - | 2 - 3 | 722 | 65 | |
| AK073161.1 | - | 2 - 3 | 575 | 65 | ||
More Information
Set Membership: |
|

LOC4329652
Genomic Map