Assay Details
| Gene Symbol: LOC4329652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770565.2 | XP_015626051.1 | 6 - 7 | 1443 | 79 | |
| GenBank mRNA | AK072252.1 | - | 6 - 7 | 1286 | 79 | |
| AK073161.1 | - | 6 - 7 | 1139 | 79 | ||
More Information
Set Membership: |
|

LOC4329652
Genomic Map