Assay Details
| Gene Symbol: LOC4329652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770565.2 | XP_015626051.1 | 7 - 8 | 1565 | 80 | |
| GenBank mRNA | AK072252.1 | - | 7 - 8 | 1408 | 80 | |
| AK073161.1 | - | 7 - 8 | 1261 | 80 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				

LOC4329652
Genomic Map