Assay Details
| Gene Symbol: LOC4330816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767757.2 | XP_015623243.1 | 1 - 2 | 591 | 116 | |
| GenBank mRNA | AK103084.1 | - | 1 - 2 | 496 | 116 | |
| DQ229362.1 | - | 1 - 2 | 436 | 116 | ||
More Information
Set Membership: |
|

LOC4330816
Genomic Map