Assay Details
| Gene Symbol: LOC4330853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771028.2 | XP_015626514.1 | 1 | 992 | 72 | |
| GenBank mRNA | AK289234.1 | - | 1 | 841 | 72 | |
More Information
Set Membership: |
|

LOC4330853
Genomic Map