Assay Details
Gene Symbol: LOC4330856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768587.2 | XP_015624073.1 | 3 - 4 | 1097 | 80 | |
GenBank mRNA | AK106777.1 | - | 1 - 2 | 2144 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
