Assay Details
Gene Symbol: LOC4330856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768587.2 | XP_015624073.1 | 4 - 5 | 1131 | 96 | |
GenBank mRNA | AK106777.1 | - | 2 - 3 | 2178 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
