Assay Details
| Gene Symbol: LOC4331528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774687.2 | XP_015630173.1 | 5 - 6 | 916 | 85 | |
| GenBank mRNA | AK067089.1 | - | 5 - 6 | 784 | 85 | |
More Information
Set Membership: |
|

LOC4331528
Genomic Map