Assay Details
Gene Symbol: LOC4331787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777261.2 | XP_015632747.1 | 4 - 5 | 885 | 102 | |
XM_015777262.2 | XP_015632748.1 | 4 - 5 | 640 | 102 | ||
GenBank mRNA | AK063978.1 | - | 4 - 5 | 543 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
